missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HO-2/HMOX2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3170-HM-010
This item is not returnable.
View return policy
Description
The Recombinant Human HO-2/HMOX2 Protein is derived from E. coli. The Recombinant Human HO-2/HMOX2 Protein has been validated for the following applications: Enzyme Activity.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl. | |
34 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
3163 | |
HO-2/HMOX2 | |
E. coli-derived human HO-2/HMOX2 protein Ser2-Leu291,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction