missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGFBP7 Protein

A cDNA sequence encoding the IGFBP7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12384-25ug

 View more versions of this product

Product Code. 15970239

  • 186.00 EUR / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

3490
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSSSDTCG PCEPASCPPL PPLGCLLGET RDACGCCPMC ARGEGEPCGG GGAGRGYCAP GMECVKSRKR RKGKAGAAAG GPGVSGVCVC KSRYPVCGSD GTTYPSGCQL RAASQRAESR GEKAITQVSK GTCEQGPSIV TPPKDIWNVT GAQVYLSCEV IGIPTPVLIW NKVKRGHYGV QRTELLPGDR DNLAIQTRGG PEKHEVTGWV LVSPLSKEDA GEYECHASNS QGQASASAKI TVVDALHEIP VKKGEGAEL
Greater than 85% as determined by SDS-PAGE.
IGFBP7 Protein
Research Use Only
IGFBP7
Human
Untagged
IGFBP7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers