missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inhibin a Protein
A cDNA sequence encoding the Inhibin a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12448-1mg
This item is not returnable.
View return policy
Specifications
Inhibin a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Alpha chain:STPLMSWPWSPSALRLLQRPPEEPAAHANCHRVALNISFQELGWERWIVYPPSFIFHYCHGGCGLHIP PNLSLPVPGAPPTPAQPYSLLPGAQPCCAALPGTMRPLHVRTTSDGGYSFKYETVPNLLTQHCACI Beta Chain:GLECDGKVNICCKKQFFVSFKDIGWNDWIIAPSGYHANYCEGECPSHIAGTSGSSLSFHSTVINHYRMRGHSPFANLKSCCVPTKLRPMSMLYYDDGQNIIKKDIQNMIVEECGCS | |
Greater than 95.0% as determined by SDS-PAGE analysis. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Inhibin-A alpha chain is supplied in 1x PBS and 50% glycerol. |