missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGALS13 Protein
A cDNA sequence encoding the LGALS13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12559-100ug
This item is not returnable.
View return policy
Specifications
LGALS13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
macglvasnl nlkpgeclrv rgevapdaks fvlnlgkdsn nlclhfnprf nahgdantiv cnskdggawg teqreavfpf qpgsvaevci tfdqanltvk lpdgyefkfp nrlnleainy maadgdfkik cvafd | |
Greater than 90.0% as determined by SDS-PAGE. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
LGALS13 protein solution (0.5 mg/ml) is formulated in 1xPBS buffer pH 7.4. |