missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LYPLA2 Protein
A cDNA sequence encoding the LYPLA2 was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 4780.00 EUR
Specifications
Gene ID (Entrez) | 11313 |
---|---|
Name | LYPLA2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LYPLA2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980969
|
enQuireBio™
QP12606-5ug |
5 μg |
120.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15970969
|
enQuireBio™
QP12606-20ug |
20 μg |
186.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15960969
|
enQuireBio™
QP12606-1mg |
1 mg |
4780.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
11313 | |
Research Use Only | |
LYPLA2 | |
Human | |
His | |
The LYPLA2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl. |
LYPLA2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MCGNTMSVPL LTDAATVSGA ERETAAVIFL HGLGDTGHSW ADALSTIRLP HVKYICPHAP RIPVTLNMKM VMPSWFDLMG LSPDAPEDEA GIKKAAENIK ALIEHEMKNG IPANRIVLGG FSQGGALSLY TALTCPHPLA GIVALSCWLP LHRAFPQAAN GSAKDLAILQ CHGELDPMVP VRFGALTAEK LRSVVTPARV QFKTYPGVMH SSCPQEMAAV KEFLEKLLPP V | |
Greater than 95.0% as determined by SDS-PAGE. |