missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human METTL21A Protein
A cDNA sequence encoding the METTL21A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12688-10ug
This item is not returnable.
View return policy
Specifications
151194 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTDRKVAL EFLKSNVQAN LPPHIQTKTV VKELTWGQNL GSFSPGEFDL ILGADIIYLE ETFTDLLQTL EHLCSNHSVI LLACRIRYER DNNFLAMLER QFTVRKVHYD PEKDVHIYEA QKRNQKEDL | |
Greater than 85% as determined by SDS-PAGE. |
METTL21A Protein | |
Research Use Only | |
METTL21A | |
Human | |
His | |
The METTL21A solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 30% glycerol. |