missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MREG Protein
A cDNA sequence encoding the MREG was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 6905.00 EUR
Specifications
Name | MREG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15981349
|
enQuireBio™
QP12727-2ug |
2 μg |
120.00 EUR
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15961349
|
enQuireBio™
QP12727-10ug |
10 μg |
186.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15971349
|
enQuireBio™
QP12727-1mg |
1 mg |
6905.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
MREG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MREG protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGLRDWL RTVCCCCGCE CLEERALPEK EPLVSDNNPY SSFGATLVRD DEKNLWSMPH DVSHTEADDD RTLYNLIVIR NQQAKDSEEW QKLNYDIHTL RQVRREVRNR WKCILEDLGF QKEADSLLSV TKLSTISDSK NTRKAREMLL KLAEETNIFP TSWELSERYL FVVDRLIALD AAEEFFKLAR RTYPKKPGVP CLADGQKELH YLPFPSP | |
Greater than 90% as determined by SDS-PAGE. |