missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUSTN1 Protein
A cDNA sequence encoding the MUSTN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12755-2ug
This item is not returnable.
View return policy
Specifications
389125 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSQAGAQ EAPIKKKRPP VKEEDLKGAR GNLTKNQEIK SKTYQVMREC EQAGSAAPSV FSRTRTGTET VFEKPKAGPT KSVFG | |
Greater than 80.0% as determined by SDS-PAGE. |
MUSTN1 Protein | |
Research Use Only | |
MUSTN1 | |
Human | |
His | |
MUSTN1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction