missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MYD88 Protein

A cDNA sequence encoding the MYD88 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12765-20ug

 View more versions of this product

Product Code. 15971469

  • 222.00 EUR / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MYD88 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMRPD RAEAPGPPAM AAGGPGAGSA APVSSTSSLP LAALNMRVRR RLSLFLNVRT QVAADWTALA EEMDFEYLEI RQLETQADPT GRLLDAWQGR PGASVGRLLE LLTKLGRDDV LLELGPSIEE DCQKYILKQQ QEEAEKPLQV AAVDSSVPRT AELAGITTLD DPLGHMPERF DAFICYCPSD IQFVQEMIRQ LEQTNYRLKL CVSDRDVLPG TCVWSIASEL IEKRCRRMVV VVSDDYLQSK ECDFQTKFAL SLSPGAHQKR LIPIKYKAMK KEFPSILRFI TVCDYTNPCT KSWFWTRLAK ALSLP
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MYD88 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.