missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYD88 Protein
A cDNA sequence encoding the MYD88 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12765-5ug
This item is not returnable.
View return policy
Specifications
MYD88 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMRPD RAEAPGPPAM AAGGPGAGSA APVSSTSSLP LAALNMRVRR RLSLFLNVRT QVAADWTALA EEMDFEYLEI RQLETQADPT GRLLDAWQGR PGASVGRLLE LLTKLGRDDV LLELGPSIEE DCQKYILKQQ QEEAEKPLQV AAVDSSVPRT AELAGITTLD DPLGHMPERF DAFICYCPSD IQFVQEMIRQ LEQTNYRLKL CVSDRDVLPG TCVWSIASEL IEKRCRRMVV VVSDDYLQSK ECDFQTKFAL SLSPGAHQKR LIPIKYKAMK KEFPSILRFI TVCDYTNPCT KSWFWTRLAK ALSLP | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MYD88 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction