Learn More
R&D Systems™ Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 6636-NX-025
346.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18186820
  • 440.00 EUR / 25µg
Explore more special offers
Description
Description
The Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera is derived from CHO. The Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
193 kDa (monomer)
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
9379
Neurexin 2 alpha/NRXN2a Isoform 2 Fc Chimera
Chinese Hamster Ovary cell line,CHO-derived human Neurexin 2 alpha/NRXN2a protein, (N-terminus) Human Neurexin 2 alpha (Met1-Thr1566) Accession &Num; NP_620060 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions