missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human POGLUT1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6437-GT-020
332.61 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to hydrolyze UDP-Glucose. The specific activity is >2 pmol/min/μg, as measured under the described conditions.Specifications
Q8NBL1 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 40-60 kDa, reducing conditions; M.W. (theoretical): 44 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
POGLUT1 | |
Recombinant | |
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 |
Enzyme Activity | |
56983 | |
Protein O-Glucosyltransferase 1/POGLUT1 | |
Chinese Hamster Ovary cell line, CHO-derived human Protein O-Glucosyltransferase 1/POGLUT1 protein Met1-Leu388, with a C-terminal 6-His tag Accession # Q8NBL1 | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane) |