missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Protocadherin-17 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 7669-CA-050

311.01 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18168120

  • 395.00 EUR / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Protocadherin-17 Protein is derived from HEK293. The Recombinant Human Protocadherin-17 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
75.1 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
27253
Protocadherin-17
Human embryonic kidney cell,HEK293-derived human Protocadherin-17 protein Leu18-Ser705,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers