missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ROR gamma/RORC/NR1F3 GST (N-Term) Protein

Recombinant Protein

597.18 EUR - 784.00 EUR

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 6097
Molecular Weight (g/mol) 37.4 kDa
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18331819
View Documents Promotion Details
Novus Biologicals™
H00006097-Q01-10ug
10 μg
597.18 EUR
10µg
Please to purchase this item. Need a web account? Register with us today!
18394529
View Documents Promotion Details
Novus Biologicals™
H00006097-Q01-25ug
25 μg
784.00 EUR
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, Protein Array, Immunoaffinity Purification
6097
Store at -80°C. Avoid freeze-thaw cycles.
RORC
>80% by SDS-PAGE and Coomassie blue staining
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
37.4 kDa
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
GPCR
ROR gamma/RORC/NR1F3
Videos
SDS
Documents

Documents

Product Certifications