missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SDHA Protein

A DNA sequence encoding the Homo sapiens (Human) SDHA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6659-ec-50ug

 View more versions of this product

Product Code. 16054880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001281261.1
SDHA Protein
Research Use Only
SDHA
Recombinant Protein
E. coli
SAKVSDSISA QYPVVDHEFD AVVVGAGGAG LRAAFGLSEA GFNTACVTKL FPTRSHTVAA QGGINAALGN MEEDNWRWHF YDTVKGSDWL GDQDAIHYMT EQAPAAVVEL ENYGMPFSRT EDGKIYQRAF GGQSLKFGKG GQAHRCCCVA DRTGHSLLHT LYGRSLRYDT SYFVEYFALD LLMENGECRG VIALCIEDGS IHRIRAKNTV VATGGYGRTY FSCTSAHTST GDGTAMITRA GLPCQDLEFV QFHPTGIYGA GCLITEGCRG EGGILINSQG ERFMERYAPV AKDLASRDVV SRSMTLEIRE GRGCGPEKDH VYLQLHHLPP EQLATRLPGI SETAMIFAGV DVTKEPIPVL PTVHYNMGGI PTNYKGQVLR HVNGQDQIVP GLYACGEAAC ASVHGANRLG ANSLLDLVVF GRACALSIEE SCRPGDKVPP IKPNAGEESV MNLDKLRFAD GSIRTSELRL SMQKSMQNHA AVFRVGSVLQ EGCGKISKLY GDLKHLKTFD RGMVWNTDLV ETLELQNLML CALQTIYGAE ARKESRGAHA REDYKVRIDE YDYSKPIQGQ QKKPFEEHWR KHTLSYVDVG TGKVTLEYRP VIDKTLNEAD CATVPPAIRS Y
Greater than 80% as determined by SDS-PAGE.
6389
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers