missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thioredoxin 80 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2329-TX-100

354.20 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18177780

  • 456.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to induce IFN-gamma secretion by human peripheral blood mononuclear cells (PBMC) in the presence of IL-2. Pekkari, K. et al. (2001) Blood 97:3184. The ED50 for this effect is 0.150-1.50 μg/mL in the presence of 8 ng/mL of recombinant human IL-2.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris, EDTA and DTT.
9 kDa (monomer)
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.50 EU / 1 μg of the protein by the LAL method.
Recombinant
7295
Thioredoxin-80
E. coli-derived human Thioredoxin-80 protein Val2-Lys81
Noncovalently-linked homodimer
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers