Learn More
R&D Systems™ Recombinant Human Vimentin Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 2105-VI-100
387.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Vimentin Protein is derived from E. coli. The Recombinant Human Vimentin Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose. | |
54.4 kDa | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
7431 | |
Vimentin | |
E. coli-derived human Vimentin protein Ser2-Glu466,with a C-terminal 6-His tag | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |