missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human ZCCHC8 GST (N-Term) Protein
Recombinant Protein
Specifications
For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
---|---|
Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 55596 |
Molecular Weight (g/mol) | 105 kDa |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
55596 | |
Store at -80°C. Avoid freeze-thaw cycles. | |
ZCCHC8 | |
ZCCHC8 |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
105 kDa | |
DKFZp434E2220, zinc finger CCHC domain-containing protein 8, zinc finger, CCHC domain containing 8 | |
>80% by SDS-PAGE and Coomassie blue staining |