missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Jilin Protein
A cDNA sequence encoding the Influenza-B Jilin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12439-10ug
This item is not returnable.
View return policy
Specifications
Influenza-B Jilin Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant B/Jilin/20/2003 solution contains 10mM Sodium phosphate, pH 7.4 and 150mM Sodium Cloride. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction