missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Measles Virus MV Large Polymerase 58-149 a. a Protein

A cDNA sequence encoding the MV Large Polymerase 58-149 a. a was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12758-500ug

 View more versions of this product

Product Code. 15971449

  • 795.00 EUR / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

MV Large Polymerase 58-149 a. a Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione, 0.1% sarcosil and 50% glycerol.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Measles Virus
Untagged
Measles Large Polymerase is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers