Learn More
R&D Systems™ Recombinant Mouse CXCL10/IP-10/CRG-2 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 466-CR-010
301.50 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse CXCL10/IP-10/CRG-2 Protein is derived from E. coli.Specifications
Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. | |
8.8 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
15945 | |
CXCL10/IP-10/CRG-2 | |
E. coli-derived mouse CXCL10/IP-10/CRG-2 protein Ile22-Pro98,with an N-terminal Met | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |