missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Endoplasmic reticulum aminopeptidase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Endoplasmic reticulum aminopeptidase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5992-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_109636.1 | |
Endoplasmic reticulum aminopeptidase 1 Protein | |
Research Use Only | |
Erap1 | |
Recombinant Protein | |
E. coli | |
PCVQRAERYF REWKSSNGNM SIPIDVTLAV FAVGAQNTEG WDFLYSKYQS SLSSTEKSQI EFSLCTSKDP EKLQWLLDQS FKGEIIKTQE FPHILTLIGR NPVGYPLAWK FLRENWNKLV QKFELGSSSI AHMVMGTTDQ FSTRARLEEV KGFFSSLKEN GSQLRCVQQT IETIEENIRW MDKNFDKIRL WLQKEKPELL | |
Greater than 80.6% as determined by SDS-PAGE. |
80898 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |