missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IFN b Protein
A cDNA sequence encoding the IFN b was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 3585.00 EUR
Specifications
Gene ID (Entrez) | 15977 |
---|---|
Name | IFN b Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Ifnb1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980209
|
enQuireBio™
QP12374-5ug |
5 μg |
120.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15970209
|
enQuireBio™
QP12374-25ug |
25 μg |
186.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15960209
|
enQuireBio™
QP12374-1mg |
1 mg |
3585.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
15977 | |
Research Use Only | |
Ifnb1 | |
Mouse | |
Untagged | |
The IFN-Beta (0.25 mg/ml) contains 20mM HEPES (pH6.0), 0.5M NaCl, 10% glycerol. |
IFN b Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MINYKQLQLQ ERTNIRKCQE LLEQLNGKIN LTYRADFKIP MEMTEKMQKS YTAFAIQEML QNVFLVFRNN FSSTGWNETI VVRLLDELHQ QTVFLKTVLE EKQEERLTWE MSSTALHLKS YYWRVQRYLK LMKYNSYAWM VVRAEIFRNF LIIRRLTRNF QN | |
Greater than 95% as determined by Analysis by SDS-PAGE. |