missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LXN Protein
A cDNA sequence encoding the LXN was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 3585.00 EUR
Specifications
Name | LXN Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970949
|
enQuireBio™
QP12599-5ug |
5 μg |
120.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15960949
|
enQuireBio™
QP12599-20ug |
20 μg |
186.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15950949
|
enQuireBio™
QP12599-1mg |
1 mg |
3585.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Spécification
LXN Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
LXN protein solution (1 mg/ml) containing Phosphate Buffer Saline (pH 7.4), 30% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEIPPTH YAASRAASVA ENCINYQQGT PHKLFLVQTV QQASKEDIPG RGHKYHLKFS VEEIIQKQVT VNCTAEVLYP QMGQGSAPEV NFTFEGEIGK NPDEEDNTFY QSLMSLKRPL EAQDIPDNFG NVSPQMKPVQ HLAWVACGYV MWQNSTEDTW YKMLKIQTVK QVQRNDDFIE LDYTILLHDI ASQEIIPWQM QVLWHPQYGT KVKHNSRLPK EGQAE | |
Greater than 95.0% as determined by SDS-PAGE. |