missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse S100A9 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 2065-S9-050
300.64 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse S100A9 Protein is derived from E. coli. The Recombinant Mouse S100A9 Protein has been validated for the following applications: Bioactivity.Specifications
P31725 | |
20202 | |
S100A9 | |
SDS-PAGE: 13kDa, reducing conditions | |
E. coli-derived mouse S100A9 protein Ala2-Lys113 | |
RUO | |
Carrier Protein | |
CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, Human S100A9 Capture, L1AG, LIAG, MAC387, MIF, MRP-14, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B) | |
Recombinant |
Lyophilized from a 0.2μm filtered solution in Tris and TCEP | |
13kDa | |
Reconstitute at 300μg/mL in Tris and TCEP. | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Ala2 | |
Noncovalently-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain |