missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Transcriptional repressor CTCFL Protein
A DNA sequence encoding the Mus musculus (Mouse) Transcriptional repressor CTCFL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5884-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_001074856.1 | |
Transcriptional repressor CTCFL Protein | |
Research Use Only | |
Ctcfl | |
Recombinant Protein | |
Yeast | |
MAAAEVPVPS GYFTQIKEQK LKPGDLEEEK EEDGVQRVEA QEGVVKEVEA ENSCLLLEAR APVESDRRIL TLQTVHLESQ DVHLQGLGWL SVPHSEELSG TVPEAEGILQ LPSVLWLDPE PQLSLQHCVT VSIPEELYPP EELQRIHFHL LRENVLMAEE NPELTPDLDE STALKKPEED EKDQLPPQGE TDKREERLLL LEMKPKEGKD DEIVLTISHL SLEEQQDPPA ANQTSVPGAK AAKPKRRRQT KGKPQSFQCD TCPFTSSKLS TFNRHIKIHS NERPHLCHLC LKAFRTVTLL RNHVNTHTGT RPHKCRDCDM AFVTSGELVR HRRYKHTYEK PFKCSLCKYA SVEASKMKRH IRSHTGERPF QCCQCAYASR DSYKLKRHMR THSGEKPYEC PTCHVRFTQS GTMKIHIAQK HGENVPKYEC PHCATIIARK SDLRVHLRNL HSQSPEEMKC RYCPAGFHER YALIQHQRTH KNEKKFKCKQ CDYACKQERC LKAHMRMHTG EKPFSCLACN KHFRQKQLLT VHLRKYHDPN FVPNLHLCLK CDKRFSRWSN LQRHRKKCDP EHETLAPNKD RRPVTRTQAS EGEAGHKEGE PQCPGEQALG HQGEAAGSQS PDHGLTCEII FNMMDK | |
Greater than 90% as determined by SDS-PAGE. |
664799 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction