missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other MOG Protein
A cDNA sequence encoding the MOG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12717-5mg
This item is not returnable.
View return policy
Specifications
MOG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The protein was lyophilized with no additives. |