missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11904-500IU
This item is not returnable.
View return policy
Specifications
FSH Protein | |
Research Use Only | |
Recombinant Protein | |
Porcine | |
MKSLQFCFLF CCWKAICCNS CELTNITITV EKEECNFCIS INTTWCAGYC YTRDLVYKDP ARPNIQKTCT FKELVYETVK VPGCAHHADS LYTYPVATEC HCGKCDSDST DCTVRGLGPS YCSFSEMKE |
500 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
The FSH was lyophilized with no additives. |