missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat BD 1 Protein

A cDNA sequence encoding the BD 1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10531-5ug

 View more versions of this product

Product Code. 15934158

  • 143.00 EUR / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

83687
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Measured by its ability to chemoattract CD34+ dendritic cells using a concentration range of 0.1-1.0 ug/ml.
Rat
Untagged
BD-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
BD 1 Protein
Research Use Only
Defb1
Recombinant Protein
E. coli
DQYRCLQNGG FCLRSSCPSH TKLQGTCKPD KPNCCRS
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.