missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque Procalcitonin Rhesus Protein
A cDNA sequence encoding the Procalcitonin Rhesus was constructed and used to recombinantly synthesize the protein.
143.00 EUR - 8190.00 EUR
Specifications
Name | Procalcitonin Rhesus Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Rhesus Macaque |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922679
|
enQuireBio™
QP13156-2ug |
2 μg |
143.00 EUR
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15902679
|
enQuireBio™
QP13156-10ug |
10 μg |
222.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15912679
|
enQuireBio™
QP13156-1mg |
1 mg |
8190.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Procalcitonin Rhesus Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rhesus Macaque | |
His | |
Procalcitonin was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in 20mM Tris buffer and 50mM NaCl, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASAPFRSALESS PDPATLSEEE ARLLLAALVQ DYVQMKASEL EQEQETEGSS LDSPRSKRCG NLSTCMLGTY TQDFNKFHTF PQTAIGVGAP GKKRDMSSDL ERNRRRYVSM PQDAN | |
Greater than 95.0% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title