missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Envelope N-BBP-His Protein

Product Code. 18680187 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18680187

Brand: Novus Biologicals™ NBP307079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation 0.2 μm filtered solution of 20 mM Tris HCl, 200 mM NaCl, pH 8.0
Gene ID (Entrez) 43740570
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias 2019-nCoV Envelope protein, COVID-19 Envelope protein, E Protein, Envelope, Human Coronavirus Envelope protein, SARSCoV2, SARS-CoV-2 E protein, SARSCoV2 Envelope protein, SARS-CoV-2 Envelope protein, Severe acute respiratory syndrome 2 Envelope Protein
Conjugate Unconjugated
Species SARS-CoV-2
Recombinant Recombinant
Form Purified
Purity or Quality Grade >95%, by reducing SDS-PAGE
Protein SARS-CoV-2 Envelope
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.