missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Yellow Fever Virus Envelope Fc Protein
930.86 EUR - 3424.32 EUR
Specifications
For Use With (Application) | ELISA |
---|---|
Formulation | DPBS pH 7.4 |
Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Gene Alias | Envelope protein E, polyprotein, partial, YFV E, YFV Envelope protein |
Purity or Quality Grade | >90% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331815
|
Novus Biologicals
NBP3-14821-0.1MG |
0.1 mg |
930.86 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18351125
|
Novus Biologicals
NBP3-14821-0.5MG |
0.5 mg |
3424.32 EUR
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
ELISA | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
>90% |
DPBS pH 7.4 | |
Envelope protein E, polyprotein, partial, YFV E, YFV Envelope protein | |
Yellow Fever Virus Envelope |