Learn More
Fisherbrand™ Rectangular Basket for Autoclaves
For use with HMT 260 FA/MA and HMT 230 FA/MA Autoclaves.
Brand:  Fisherbrand™ 52037
 View more versions of this product
Product Code. 15608835
  • 133.00 EUR / Each
Estimated Shipment: 09-05-2025
Description
  • Solid construction
  • Wire mesh basket
  • 370 x 210 x 105mm or 340 x 180 x 120mm
Specifications
Specifications
Autoclavable
Basket, Rectangular
370 x 210 x 105 mm
HMT 260 FA/MA Tabletop Autoclaves
Wire Mesh