Learn More
Hach™ Rectangular PMMA Sample Cells
Hach™ Rectangular PMMA Sample Cells are acrylic semi-micro cuvettes with caps that measure 50mmH x 19.5mmW x 60mmD.
Brand: Hach™ LZP341
This item is not returnable.
View return policy
Description
Ten 7mL sample cells per pack.
Order Info
dimProductType
Specifications
Clear | |
Caps | |
7 mL | |
Sample Cell |
PMMA (acrylic glass) | |
10 Each | |
50mmH x 19.5mmW x 60mmD |