Learn More
Fisherbrand™ PTFE Stir Bar Retriever
Use to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Alnico V magnets at its core.
Brand: Fisherbrand™ 004.450
Description
- Inert and leak-free
- Molded in pure, FDA-approved PTFE
- Alnico V magnets
- Autoclavable
Specifications
White | |
Removal of Stirrer Bars from vessels | |
10mm | |
450 mm |
PTFE encapsulated Alnico V magnet core | |
PTFE Stirrer Bars | |
10 mm | |
1/Ea. |
Product Content Correction
Product Title
Fisherbrand™ PTFE Stir Bar Retriever > Dimensions (Dia. x L): 10 dia.x 450mmL
Spot an opportunity for improvement?Share a Content Correction