missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl

SI Analytics™ Redox Solution, 430mV Pt/Calomel, 470mV Pt/Ag/AgCl is designed to be used in redox tests.

Brand:  SI Analytics™ 285138168


 View more versions of this product

Product Code. 11964739

  • 50.40 EUR / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • GHS07: Skin Irrit. 2 H315 Causes skin irritation, Eye Irrit. 2 H319 Causes serious eye irritation
  • Precautionary statements: P280 Wear protective gloves / eye protection / face protection. P264 Wash thoroughly after handling. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P321 Specific treatment (see on this label). P332+P313 If skin irritation occurs: Get medical advice/attention. P337+P313 If eye irritation persists: Get medical advice/attention. P302+P352 IF ON SKIN: Wash with plenty of water.
  • Ingredients with limit values that require monitoring at the workplace: 7664-93-9 sulphuric acid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

102°C
430/470 Pt/Kalomel, 470 Pt/Ag/AgclMv
Odorless
-0.1
Fully miscible in water
Light Yellow
Redox
DURAN™ Bottle
250 mL
23 hPa
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H315-Causes skin irritation.,Causes skin irritation.
  • H290-May be corrosive to metals.
  • H319-Causes serious eye irritation.,Causes serious eye irritation.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.,IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers