Learn More
BINDER™ Reinforced Heating Output
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers FED 720
Unit Size:
Each
Description
- 400 V option model
- Heats up faster to temperatures up to 300°C
- Improves recovery time
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers FED 720 |
| Temperature (Metric) | 300°C (Max.) |
| Type | Heating Output |
| For Use With (Application) | For Faster Heating up Time and Improved Recovery Time |
| Voltage | 400 V |