Learn More
Thermo Scientific™ Remel™ Gram Crystal Violet
Product Code. p-4525083
Click to view available options
Quantity:
5/Pk.
Each
Unit Size:
250mL
4 L
Pack of 5
Product Code. 11982952
Brand: Thermo Scientific™ R40052
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY
Specifications
Type Crystal Violet
Quantity Each
Certifications/Compliance CE Marked
Volume (Metric) 250 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.