Learn More
BRAND™ Replacement PFA Screw Cap
Shop All Brand Products
Click to view available options
Closure Size:
GL 18
GL 25
Unit Size:
Each
Description
- Constructed of PFA
- For use with volumetric flasks
- Replacement
Specifications
Specifications
| Material | PFA |
| For Use With (Equipment) | Volumetric Flask |
| Height (Metric) | 2.2 cm |
| Length (Metric) | 8 cm |
| Product Type | Screw cap |
| Width (Metric) | 3.4 cm |
| Closure Size | GL 25 |