missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Replica Plating Apparatus
Replica Plating Apparatus for safely and easily spreading yeast and bacterial cells on to the plate in seconds.
Brand: MP Biomedicals™ 115000001
This item is not returnable.
View return policy
Description
- Wooden sphere with block of wood with hole for sphere
- Safely and easily spread yeast and bacterial cells
- Spread cells in just seconds per plate
- Eliminate hazards of flame and ethanol
- No need for fragile glass ''hockey sticks''
- Consistent, even colony distribution
- Sterile and easy to use
- Made from hardwood
- Can be used with velvet, gauze, or even paper towels to transfer colonies from plate to plate or plate to filter while retaining colony orientation
- Replica Plating Apparatus and Velvet Squares can be ordered to fit either 100 mm or 150 mm plates
- Applications: Bacteria Cell Culture, Cell Preservation, Cell Counting, Viability, and Cryopreservation, Insect Cell Culture, Mammalian Cell Culture, Plant Cellgrowth, Primary Cell Culture, Stem Cell Culture, Yeast Cell Culture
- Store at room temperature
Specifications
Room Temperature | |
Hardwood | |
100 mm |
Wooden Sphere with Block of wood with Hole for Sphere | |
Bacteria Cell Culture, Cell Preservation, Cell Counting, Viability, and Cryopreservation, Insect Cell Culture, Mammalian Cell Culture, Plant Cellgrowth, Primary Cell Culture, Stem Cell Culture, Yeast Cell Culture | |
100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction