Learn More
Brady™ Repositionable Multipurpose Vinyl Cloth Labels
Click to view available options
Dimensions (LxW):
1 x 1.9 in.
12.70 mm (W) x 12.70 mm (H)
19.05 mm (W) x 12.70 mm (H)
25.40 mm (W) x 12.70 mm (H)
25.40 mm (W) x 19.05 mm (H)
25.40 mm (W) x 25.40 mm (H)
25.40 mm (W) x 31.75 mm (H)
25.40 mm (W) x 38.10 mm (H)
6.10 mm (W) x 4.88 m (L)
8.08 mm (W) x 4.88 m (L)
9.53 mm (W) x 6.10 m (L)
For Use With (Equipment):
BMP41, BMP51, M511
BMP41;BMP51;BMP53;M511;M410;M510
BMP51;BMP53;M511;M510
Format:
Continuous
Die-cut
Unit Size:
Each
Pack of 100
Pack of 120
Pack of 160
Pack of 180
Pack of 220
Pack of 240
Beschreibung
- Ideal for marking wires, cables, terminal blocks, wall plates
- Constructed of tough coated vinyl cloth for resistance to extreme temperatures
- Made with a strong rubber-based adhesive that can be cleanly removed and securely replaced on any clean, dry surface
Spezifikation
Spezifikation
| Certifications/Compliance | UL Recognised Compliant with Directive 2011/65/EU and 2015/863 |
| Adhesive Type | Repositionable Rubber |
| Color | White |
| Finish | Semi-gloss |
| Format | Continuous |
| Material | Vinyl |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Asset tracking;Barcode labelling;Circuit board labelling;Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;General identification;Wire and cable labelling |
| For Use With (Equipment) | BMP41;BMP51;BMP53;M511;M410;M510 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur