missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ and SOURCE™ 15 and 5RPC columns

High Performance Reversed Phase Chromatography

434.00 EUR - 1170.00 EUR

Specifications

Ligand Type Matrix surface
Particle Shape Monosized
Particle Size 15 μm
pH 1 to 12
Packing Material Polystyrene/Divinyl Benzene
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11580794
View Documents
Cytiva
17-0727-20
10 mL
434.00 EUR
Pack of 10
In Stock
to see stock.
Add to basket
 
10515595
View Documents
Cytiva
17-5068-01
1.7 mL
1170.00 EUR
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Ideally suited for difficult preparative separations at all scales

  • High resolution separations in minutes
  • High performance at low back pressures
  • High capacity at high flow rates
  • Wide pH stability range
  • Excellent scalability
  • Reproducible quality

Specifications

Specifications

Matrix surface
15 μm
Polystyrene/Divinyl Benzene
Reversed Phase Column
Monosized
1 to 12
No
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SOURCE 15RPC, 10 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.