missing translation for 'onlineSavingsMsg'
Learn More

Resveratrol from Synthetic, ≥98%, MP Biomedicals™

Product Code. 11482681
Click to view available options
Quantity:
0.1 mg
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 501-36-0
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00133799
InChI Key LUKBXSAWLPMMSZ-OWOJBTEDSA-N
Synonym resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
PubChem CID 445154
ChEBI CHEBI:45713
IUPAC Name 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 25 mg
Formula Weight 228.25
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Resveratrol, from Synthetic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.