missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Reticle, Crosshair, 19mm dia.
Plain crosshair-type eyepiece reticle. Motic™ Reticle, Crosshair, 25mm dia. divides the viewing field into 4 even quadrants.
Brand: Motic™ 12180145-R
This item is not returnable.
View return policy
Description
- Evenly divides the viewing field into 4 quadrants with a simple crosshair.
- Not graduated for measuring.
Specifications
Simple Crosshair | |
19 mm | |
Eyepiece, 19mm dia., PM-Series Microscopes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction