Learn More
Fisherbrand™ Reusable Dispenser Tips
Robust, stable, and durable dispenser tips for several brands of pipettors.
Brand:  Fisherbrand™ FB56378
Product Code. 11542363
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
These tips are for use with the following pipettors:
  • Brand Handystep™ and Handystep™ electronic All tips
  • Eppendorf™ Multipette™ (not Multipette™ Plus) All tips
  • Gilson Distriman™ 
  • Ritter Ripette™ All tips
  • Tri-continent Easystep™ 
Additonal features:
  • These Dispenser™ tips are non-sterile and available by gamma irradiation
  • These dispenser tips are re-useable
  • The adapter for this dispenser tip is autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With one autoclavable adapter
50 mL
Brand Handystep and Handystep, Electronic Eppendorf, Multipette (not Multipette Plus), Gilson Distriman, Ritter Ripette. Tri-continent Easystep
25/Pk.
Dispenser Tip