Learn More
3M™ Reusable Half Face Mask Respirator
3M™ Reusable Half Mask has been designed with user comfort and convenience in mind. Compatible with DIN threaded, 3M™ Filters DT-Series, the HF-301 offers protection against a wide range of respiratory hazards.
Brand: 3M™ 7100265229
313.97 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hybrid reflex seal contours and flexes to the user's face to aid wearer comfort
- Seal check mechanism - when pressed during exhale, the mask inflates slightly showing that a good seal has been achieved
- Integrated voice panel for enhanced voice intelligibility while wearing the mask
- Nose pinch allows for improved integration with eyewear
- Comfortable, durable, low profile head cradle & thick elasticated straps with simple rear fastening
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Half Facepiece Respirator | |
DIN Threaded, 3M™ Filters DT-Series | |
10/Cs. |
Large | |
HF-303 |