Learn More
3M™ Reusable Half Mask Respirator Kit A2P3 R
3M™ Reusable Half Mask Respirator Kit A2P3 R is a safe solution for common applications in industries such as agriculture, construction, and painting.
Brand: 3M™ 7000061553
This item is not returnable.
View return policy
Description
- Ready to use respiratory kit with 3M™ Reusable Half Face Mask 7503L or 7503M and 3M™ Filters offering A2P3 R protection
- Half mask with a soft silicone face piece for comfort
- Bayonet connection fitting system to easily connect filters to mask
- 3M™ Cool Flow™ Valve provides easier breathing and reduced heat and moisture build-up
- Low profile, twin filter design half mask for a balanced secure fit
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CE Approved | |
Half Mask Respirator Kit | |
Half mask and filter |
A2P3 R | |
Medium |