Learn More
3M™ Reusable Half Mask Respirator Kit A2P3 R
3M™ Reusable Half Mask Respirator Kit A2P3 R is a safe solution for common applications in industries such as agriculture, construction, and painting.
Brand:  3M™ 7000061555
 View more versions of this product
Product Code. 15779797
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Ready to use respiratory kit with 3M™ Reusable Half Face Mask 7503L or 7503M and 3M™ Filters offering A2P3 R protection
  • Half mask with a soft silicone face piece for comfort
  • Bayonet connection fitting system to easily connect filters to mask
  • 3M™ Cool Flow™ Valve provides easier breathing and reduced heat and moisture build-up
  • Low profile, twin filter design half mask for a balanced secure fit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Approved
Half Mask Respirator Kit
Half mask and filter
A2P3 R
Large