missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOBTB2 Mouse anti-Human, Clone: DBC2/3361, Novus Biologicals™
Mouse Monoclonal Antibody
242.97 EUR - 483.79 EUR
Specifications
Antigen | RHOBTB2 |
---|---|
Clone | DBC2/3361 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18384623
|
Novus Biologicals
NBP3-13899-20UG |
20 μg |
242.97 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331667
|
Novus Biologicals
NBP3-13899-100UG |
100 μg |
483.79 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
RHOBTB2 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Signal Transduction | |
10 mM PBS with 0.05% BSA | |
DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2 | |
Human recombinant RHOBTB2 protein fragment (around aa 554-604) (exact sequence is proprietary) (Uniprot: Q9BYZ6) | |
Primary | |
Store at 4°C. |
DBC2/3361 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9BYZ6 | |
23221 | |
IgG2b κ | |
Protein A or G purified |