missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodium Potassium Chloride, K2RhCl5, MP Biomedicals
Click to view available options
Quantity:
1 g
Specifications
Specifications
Quantity | 1 g |
Formula Weight | 358.37 |
Chemical Name or Material | RHODIUM POTASSIUM CHLORIDE |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'